Skip to Content
Merck
All Photos(1)

Key Documents

AMBH95E0826C

Ambeed, Inc.

3-Pyridinemethanethiol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
¥32,500
250 MG
¥55,500
1 G
¥118,600
5 G
¥271,200

Vendor SKU: A379276-100MG

¥32,500


Ships in 12 days - from Aldrich Partner.


Select a Size

Change View
100 MG
¥32,500
250 MG
¥55,500
1 G
¥118,600
5 G
¥271,200

About This Item

CAS Number:
Molecular Weight:
125.19
MDL number:

Vendor SKU: A379276-100MG

¥32,500


Ships in 12 days - from Aldrich Partner.

Iupac Name

3-pyridinylmethanethiol

Inchi Code

1S/C6H7NS/c8-5-6-2-1-3-7-4-6/h1-4,8H,5H2

InChI key

MBXKCLHOVPXMCJ-UHFFFAOYSA-N

storage temp.

Inert atmosphere,2-8C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.ambeed.com/products/17617-05-9.html

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

MSDS

https://www.ambeed.com/products/17617-05-9.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service