Skip to Content
Merck
All Photos(1)

Key Documents

AMBH303C44AA

Ambeed, Inc.

α-D-Mannose pentaacetate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
¥10,900
5 G
¥11,500
10 G
¥12,400
25 G
¥13,300
100 G
¥24,300

Vendor SKU: A116304-1G

¥10,900


Ships in 12 days - from Aldrich Partner.


Select a Size

Change View
1 G
¥10,900
5 G
¥11,500
10 G
¥12,400
25 G
¥13,300
100 G
¥24,300

About This Item

CAS Number:
Molecular Weight:
390.34
MDL number:

Vendor SKU: A116304-1G

¥10,900


Ships in 12 days - from Aldrich Partner.

Iupac Name

1,2,3,4,6-penta-O-acetyl-alpha-D-mannopyranose

Inchi Code

1S/C16H22O11/c1-7(17)22-6-12-13(23-8(2)18)14(24-9(3)19)15(25-10(4)20)16(27-12)26-11(5)21/h12-16H,6H2,1-5H3/t12-,13-,14+,15+,16+/m1/s1

InChI key

LPTITAGPBXDDGR-OWYFMNJBSA-N

storage temp.

Sealed in dry,Store in freezer, under -20C

Purity

98%

Country of Origin

CN

Shipping Temperature

Shipped in cold pack

Physical Form

solid

Product Link

https://www.ambeed.com/products/4163-65-9.html

Synonym

(2R,3S,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service