Skip to Content
Merck
All Photos(1)

Key Documents

AMBH2D6F40E3

Ambeed, Inc.

3-Carbamoylbenzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
¥15,300
250 MG
¥17,900
1 G
¥20,100
5 G
¥44,400

Vendor SKU: A111256-100MG

¥15,300


Ships in 12 days - from Aldrich Partner.


Select a Size

Change View
100 MG
¥15,300
250 MG
¥17,900
1 G
¥20,100
5 G
¥44,400

About This Item

CAS Number:
Molecular Weight:
165.15
MDL number:

Vendor SKU: A111256-100MG

¥15,300


Ships in 12 days - from Aldrich Partner.

Iupac Name

3-(aminocarbonyl)benzoic acid

Inchi Code

1S/C8H7NO3/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4H,(H2,9,10)(H,11,12)

InChI key

FVUKYCZRWSQGAS-UHFFFAOYSA-N

storage temp.

Sealed in dry,Room Temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/4481-28-1.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service