Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

G1652

Sigma-Aldrich

Gold colloid

~0.01% HAuCl4, ~1 A520 units/mL, 17-23 nm mean particle size (monodisperse), 20 nm particle size

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 295.20
5 G
CHF 763.80

Vendor SKU: PC303516-1G

CHF 295.20


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
CHF 295.20
5 G
CHF 763.80

About This Item

Empirical Formula (Hill Notation):
Au
CAS Number:
Molecular Weight:
196.97
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Vendor SKU: PC303516-1G

CHF 295.20


Ships in 14 days - from Aldrich Partner.

description

<0.01% tannic acid
<0.04% sodium citrate

form

dispersion
nanoparticles

contains

0.02% sodium azide as preservative

concentration

~0.01% HAuCl4
~1 A520 units/mL

mean particle size

17-23 nm (monodisperse)

particle size

20 nm

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

See "Immunocytochemical Methods and Protocols" Lorette C. Javois, chapter 8 (Sigma product I5653) for microscopy related applications.

Suitability

Suitable for adsorption of biomolecules for labeled probe synthesis.

Physical form

Colloidal suspension

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

G1652-100ML:
G1652-25ML:
G1652-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service