Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

81535

Sigma-Aldrich

Polystyrene A-Br

suitable for peptide synthesis, extent of labeling: 0.8-1.2 mmol/g Br loading

Synonym(s):

(2-Bromoethyl)polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis
reactivity: nucleophile reactive

extent of labeling

0.8-1.2 mmol/g Br loading

matrix

crosslinked with 1% DVB

particle size

500-560 μm

application(s)

peptide synthesis

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Brominated polystyrene is a core matrix for the preparation of various functionalized polystyrene resins for solid phase peptide synthesis (SPPS) and solid phase organic synthesis (SPOS).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

81535-VAR:
81535-BULK:
81535-25G:
81535-5G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service