Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

701548

Sigma-Aldrich

4-Fluorophenylboronic acid MIDA ester

97%

Synonym(s):

2-(4-Fluorophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11BFNO4
CAS Number:
Molecular Weight:
251.02
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

231-236 °C

functional group

fluoro

SMILES string

CN1CC(=O)OB(OC(=O)C1)c2ccc(F)cc2

InChI

1S/C11H11BFNO4/c1-14-6-10(15)17-12(18-11(16)7-14)8-2-4-9(13)5-3-8/h2-5H,6-7H2,1H3

InChI key

VRWBUFHGNOFGKX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

701548-BULK:
701548-5G:
701548-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service