Skip to Content
Merck
All Photos(1)

Documents

Safety Information

631213

Sigma-Aldrich

3-[5-(4-Chlorophenyl)furan-2-yl]acrylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9CIO3
CAS Number:
Molecular Weight:
352.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

195-199 °C (lit.)

SMILES string

OC(=O)\C=C\c1ccc(o1)-c2ccc(Cl)cc2

InChI

1S/C13H9ClO3/c14-10-3-1-9(2-4-10)12-7-5-11(17-12)6-8-13(15)16/h1-8H,(H,15,16)/b8-6+

InChI key

FPSINWFYJYHUBV-SOFGYWHQSA-N

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

631213-1G:
631213-BULK:
631213-5G:
631213-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service