Skip to Content
Merck
All Photos(1)

Documents

Safety Information

451185

Sigma-Aldrich

Neodymium(III) carbonate hydrate

99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Nd2(CO3)3 · xH2O
CAS Number:
Molecular Weight:
468.51 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: neodymium

mp

114 °C (lit.)

SMILES string

O.[Nd+3].[Nd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

InChI

1S/3CH2O3.2Nd.H2O/c3*2-1(3)4;;;/h3*(H2,2,3,4);;;1H2/q;;;2*+3;/p-6

InChI key

CZXBEKQIOBCHSG-UHFFFAOYSA-H

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

451185-BULK:
451185-100G:
451185-25G:
451185-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service