Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

400734

Sigma-Aldrich

Mercury(II) sulfide red

synthesis grade

Synonym(s):

Mercuric sulfide red

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HgS
CAS Number:
Molecular Weight:
232.66
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

synthesis grade

color

black

mp

583.5 °C (lit.)

density

7.6 g/mL at 25 °C

SMILES string

S=[Hg]

InChI

1S/Hg.S

InChI key

QXKXDIKCIPXUPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

400734-250G:
400734-BULK:
400734-VAR:
400734-50G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masaki Ohata et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 31(8), 855-858 (2015-08-11)
The effect of long-time X-ray irradiation on Cr and Hg in a polypropylene disk certified reference material (PP disk CRM, NMIJ CRM 8136-a) during measurements by X-ray fluorescence (XRF) spectrometry was examined in the present study. The XRF intensities of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service