Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

381365

Sigma-Aldrich

Germanium(IV) bromide

Synonym(s):

Germanium tetrabromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GeBr4
CAS Number:
Molecular Weight:
392.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

reaction suitability

reagent type: catalyst
core: germanium

bp

186.5 °C (lit.)

mp

26.1 °C (lit.)

density

3.13 g/mL at 25 °C (lit.)

SMILES string

Br[Ge](Br)(Br)Br

InChI

1S/Br4Ge/c1-5(2,3)4

InChI key

VJHDVMPJLLGYBL-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

381365-10G:
381365-VAR:
381365-BULK:
381365-10G-PW:
381365-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service