Skip to Content
Merck
All Photos(1)

Documents

345954

Sigma-Aldrich

N-(3-Indolylacetyl)-L-phenylalanine

99%

Synonym(s):

IAA-L-Phe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N2O3
CAS Number:
Molecular Weight:
322.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

99%

optical activity

[α]20/D +36°, c = 3 in ethanol

mp

154-156 °C (lit.)

SMILES string

OC(=O)[C@H](Cc1ccccc1)NC(=O)Cc2c[nH]c3ccccc23

InChI

1S/C19H18N2O3/c22-18(11-14-12-20-16-9-5-4-8-15(14)16)21-17(19(23)24)10-13-6-2-1-3-7-13/h1-9,12,17,20H,10-11H2,(H,21,22)(H,23,24)/t17-/m0/s1

InChI key

BUGQHORRADGONS-KRWDZBQOSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service