Skip to Content
Merck
All Photos(2)

Documents

Safety Information

211419

Sigma-Aldrich

5-Bromo-1,2,4-trimethylbenzene

99%

Synonym(s):

5-Bromopseudocumene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2(CH3)3
CAS Number:
Molecular Weight:
199.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

233-235 °C (lit.)

mp

71-73 °C (lit.)

SMILES string

Cc1cc(C)c(Br)cc1C

InChI

1S/C9H11Br/c1-6-4-8(3)9(10)5-7(6)2/h4-5H,1-3H3

InChI key

SCZXFZRJDVZMJI-UHFFFAOYSA-N

Application

5-Bromo-1,2,4-trimethylbenzene was used in the preparation of:
  • 1′,2′-dideoxy-1′-(1,2,4-trimethylphenyl)-3′,5′-di-O-toluoyl-β-D-ribofuranose
  • ubiquinones, also referred as Coenzyme Qn
  • 2, 4, 5-trimethylbenzoic acid

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

211419-25G:
211419-BULK:
211419-100G:
211419-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Ubiquinones Utilizing Pd (0)-Catalyzed Stille Coupling.
Jung YS, et al.
Bull. Korean Chem. Soc., 21(5), 463-464 (2000)
2, 4, 5-Trimethylbenzoic Acid.
Barcon A, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 53(12), 1842-1845 (1997)
Barbara A Schweitzer et al.
The Journal of organic chemistry, 59(24), 7238-7242 (1994-12-01)
Described are the design, synthesis, and structures of three nonpolar nucleoside isosteres to be used as probes of noncovalent bonding in DNA and as isosteric replacements for the natural nucleosides in designed nucleic acid structures. Reaction of substituted aryl Grignards

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service