Skip to Content
Merck
All Photos(1)

Key Documents

AC7448879813

Advanced Chemical Intermediates Ltd.

4-[1-Hydroxy-ethylidene]-2-phenyl-4H-oxazol-5-one Mix of E&Z isomers

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$158.70
500 MG
$226.55
1 G
$317.40
5 G
$1,135.05
10 G
$1,928.55
25 G
$3,790.40

Vendor SKU: 15541-250MG

$158.70


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$158.70
500 MG
$226.55
1 G
$317.40
5 G
$1,135.05
10 G
$1,928.55
25 G
$3,790.40

About This Item

Molecular Weight:
203.2
MDL number:

Vendor SKU: 15541-250MG

$158.70


Ships in 7 days - from Aldrich Partner.

Iupac Name

(4E)-4-(1-hydroxyethylidene)-2-phenyl-1,3-oxazol-5(4H)-one

Inchi Code

1S/C11H9NO3/c1-7(13)9-11(14)15-10(12-9)8-5-3-2-4-6-8/h2-6,14H,1H3

InChI key

HQKJROKUOYLELW-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Room temperature

Physical Form

Beige solid

Product Link

https://www.acints.com/products/SB-08K-003

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.acints.com/products/SB-08K-003


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service