AC7372907640
Advanced Chemical Intermediates Ltd.
Ethyl 2-(2-bromobenzyl)thiazole-4-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Molecular Weight:
326.21
MDL number:
Iupac Name
ethyl 2-(2-bromobenzyl)-1,3-thiazole-4-carboxylate
Inchi Code
1S/C13H12BrNO2S/c1-2-17-13(16)11-8-18-12(15-11)7-9-5-3-4-6-10(9)14/h3-6,8H,2,7H2,1H3
InChI key
AVNINJXMPZETJD-UHFFFAOYSA-N
storage temp.
Room temperature
Purity
95%
Country of Origin
GB
Shipping Temperature
Room temperature
Product Link
https://www.acints.com/products/SB-02D-003
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.acints.com/products/SB-02D-003
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service