Skip to Content
Merck
All Photos(1)

Key Documents

AC5H0A31374A

Accela ChemBio Inc.

Allyl 2-Oxocyclopentanecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
168.19
MDL number:

Iupac Name

allyl 2-oxocyclopentane-1-carboxylate

Inchi Code

1S/C9H12O3/c1-2-6-12-9(11)7-4-3-5-8(7)10/h2,7H,1,3-6H2

InChI key

JBVYBVWYOSXHFO-UHFFFAOYSA-N

storage temp.

+4C

Purity

95%

Country of Origin

CN

Shipping Temperature

+4C

Physical Form

Colorless to red liquid

Product Link

http://www.accelachem.com/cn/search.html?keyword=SY224713&attname=GoodsCode

MSDS

http://www.accelachem.com/cn/msds/SY224713.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service