Skip to Content
Merck
All Photos(1)

Key Documents

AABH97D28E30

AA BLOCKS, INC.

2-Bromo-1-fluoro-4-(prop-2-yn-1-yloxy)benzene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
¥250,400

Vendor SKU: AA00OQ5J-5G

¥250,400


Ships in 26 days - from Aldrich Partner.


Select a Size

Change View
5 G
¥250,400

About This Item

CAS Number:
Molecular Weight:
229.05
MDL number:

Vendor SKU: AA00OQ5J-5G

¥250,400


Ships in 26 days - from Aldrich Partner.

Iupac Name

2-bromo-1-fluoro-4-(prop-2-yn-1-yloxy)benzene

Inchi Code

1S/C9H6BrFO/c1-2-5-12-7-3-4-9(11)8(10)6-7/h1,3-4,6H,5H2

InChI key

PKPUZAGUCCRHQX-UHFFFAOYSA-N

storage temp.

2~8C

Purity

95%

Country of Origin

CN

Product Link

https://www.aablocks.com/prod/1548615-39-9

Synonym

2-Bromo-1-fluoro-4-(prop-2-yn-1-yloxy)benzene

MSDS

https://www.aablocks.com/downloadSds?cas=1548615-39-9


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service