Skip to Content
Merck
All Photos(1)

Key Documents

AABH97D200A6

AA BLOCKS, INC.

2(1H)-Quinolinone, 7-(trifluoromethyl)-

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
¥141,900
1 G
¥233,100

Vendor SKU: AA0003RX-250MG

¥141,900


Ships in 26 days - from Aldrich Partner.


Select a Size

Change View
250 MG
¥141,900
1 G
¥233,100

About This Item

CAS Number:
Molecular Weight:
213.16
MDL number:

Vendor SKU: AA0003RX-250MG

¥141,900


Ships in 26 days - from Aldrich Partner.

Iupac Name

7-(trifluoromethyl)quinolin-2(1H)-one

Inchi Code

1S/C10H6F3NO/c11-10(12,13)7-3-1-6-2-4-9(15)14-8(6)5-7/h1-5H,(H,14,15)

InChI key

STHFBPRDYLWJGX-UHFFFAOYSA-N

storage temp.

RT

Country of Origin

US

Product Link

https://www.aablocks.com/prod/1011533-24-6

Synonym

2-Hydroxy-7-(trifluoromethyl)quinoline

MSDS

https://www.aablocks.com/downloadSds?cas=1011533-24-6


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service