GC Analysis of a 37-Component FAME Mix on SP™-2560
Conditions | |
---|---|
column | SP-2560, 100 m x 0.25 mm I.D., 0.20 µm (24056) |
oven | 140 °C (5 min), 8 °C/min to 180 °C, 4 °C/min to 210 °C, 20 °C/min to 250 °C (7 min) |
carrier gas | hydrogen, 40 cm/sec |
sample | Supelco 37-Component FAME Mix (47885-U), analytes at concentrations indicated in methylene chloride |
injection | 1 µL, 100:1 split |
liner | 4 mm I.D., cup design |
inj. temp. | 250 °C |
detector | FID, 250 °C |
Description | |
---|---|
Categories | Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs) |
Featured Industry | Agriculture Clinical Food and Beverages |
Legal Information | SP is a trademark of Sigma-Aldrich Co. LLC |
Other Notes | Complementary products: Syringes Vials Gas Purification and Gas Management Inlet Liners, Glass Wool, and Specialized Hand Tools Septa and Specialized Hand Tools Column Ferrules, Nuts, and Specialized Hand Tools |
suitability | application for GC |
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch