GC Analysis of FAMEs in B20 Biodiesel (Soy Source) on Omegawax
CONDITIONS
column
Omegawax 250, 30 m x 0.25 mm I.D., 0.25 μm (24136)
oven
50 °C, 13 °C/min to 270 °C (5 min)
inj. temp.
250 °C
detector
FID, 270 °C
detector
helium, 40 cm/sec
injection
1 μL 100:1 split
liner
4 mm I.D. FocusLiner™ inlet liner (no taper)
sample
B20 biodiesel (soy source) diluted 1:20 in n-hexane
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
null