Merck
  • Epoxy resin as a new alternative sorbent phase for stir bar sorptive extraction for the determination of triclosan and methyl-triclosan.
Epoxy resin as a new alternative sorbent phase for stir bar sorptive extraction for the determination of triclosan and methyl-triclosan.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2017-08-02)
Jackeline Stoski, Natalicio F Leite, Ronei E da Paixão, Gilberto Abate
ABSTRACT
The main objective of this study is to propose a new sorbent phase based on a commercial epoxy resin, for use as an alternative sorbent in the stir bar sorptive extraction (SBSE) technique. The analytes triclosan (TCS) and methyl-triclosan (MTCS) were selected in order to demonstrate the application of the bars, using some water samples as matrices. The extraction conditions of sample volume, stirring time, pH, temperature and ionic strength were evaluated, and for the back extraction, the bars were sonicated using 1.00 mL of acetonitrile for 10 min. The technique of liquid chromatography using a diode array detector was employed for the quantitation. Analytical curves of between 2.50 and 50.0 μg L
Product Description
Supelco
Triclosan-methyl, PESTANAL®, analytical standard
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Octadecylsilane, 97%