Merck
  • The Expanding Family of Natural Anion Channelrhodopsins Reveals Large Variations in Kinetics, Conductance, and Spectral Sensitivity.
The Expanding Family of Natural Anion Channelrhodopsins Reveals Large Variations in Kinetics, Conductance, and Spectral Sensitivity.
Scientific reports (2017-03-04)
Elena G Govorunova, Oleg A Sineshchekov, Elsa M Rodarte, Roger Janz, Olivier Morelle, Michael Melkonian, Gane K-S Wong, John L Spudich
ABSTRACT
Natural anion channelrhodopsins (ACRs) discovered in the cryptophyte alga Guillardia theta generate large hyperpolarizing currents at membrane potentials above the Nernst equilibrium potential for Cl
Sigma-Aldrich
all trans-Retinal, powder, ≥98%