Merck
  • Anion recognition and redox sensing amplification by self-assembled monolayers of 1,1'bis(alkyl-N-amido)ferrocene.
Anion recognition and redox sensing amplification by self-assembled monolayers of 1,1'bis(alkyl-N-amido)ferrocene.
Chemical communications (Cambridge, England) (2002-08-29)
Paul D Beer, Jason J Davis, Daniel A Drillsma-Milgrom, Fridrich Szemes
ABSTRACT
Surface pre-organisation within robust amidoferrocene self-assembled monolayers can be exploited in the selective electrochemical sensing of anions in both organic and aqueous media.