Merck
  • Synthesis of 2,5-Diaryl-1,5-dienes from Allylic Bromides Using Visible-Light Photoredox Catalysis.
Synthesis of 2,5-Diaryl-1,5-dienes from Allylic Bromides Using Visible-Light Photoredox Catalysis.
The Journal of organic chemistry (2015-10-31)
Gerald Pratsch, Larry E Overman
ABSTRACT
Visible-light photoreductive coupling of 2-arylallyl bromides in the presence of the photocatalyst Ru(bpy)3(PF6)2, a Hantzsch ester, and i-Pr2NEt gives 2,5-diaryl-1,5-dienes in high yield. This method avoids the use of stoichiometric metal reductants and is compatible with the presence of halogen, alkyl, electron-donating, and electron-withdrawing substituents on the aromatic ring.
Sigma-Aldrich
2,3-Dibromopropene, 80%, technical grade