Merck
  • Prolonged febrile seizures, clinical characteristics, and acute management.
Prolonged febrile seizures, clinical characteristics, and acute management.
Epilepsia (2013-04-05)
Haim Bassan, Marina Barzilay, Shlomo Shinnar, Zamir Shorer, Israel Matoth, Varda Gross-Tsur
ABSTRACT
Prolonged febrile seizures (PFS) lasting ≥15 min have been associated with increased risk for epilepsy in later life. Initial treatment, mostly prehospital, aims to prevent its evolution to febrile status epilepticus (FSE) and reduce adverse outcome. Paucity of information is available on the immediate treatment before reaching a hospital facility. We obtained data, prospectively, on all children who presented from January 2008 to March 2010 with PFS to the emergency rooms of four Israeli medical centers. Information related to seizure semiology, treatment, and medical history was collected into a predefined pro forma form and reviewed centrally. Sixty children, median age 18.3 months (interquartile range [IQR] 12-28) were included with a median seizure duration of 35 min (IQR 26-60), 43 (71.7%) lasting ≥30 min. Seizures had focal onset in 34 infants (57%). Fifty-four families (90%) activated the ambulance service; median ambulance arrival time was 8 min (IQR 5-10), 33 (61%) were medically treated by the ambulance paramedic, of whom 15 (45%) responded to treatment. Twelve children with active seizures did not receive medications. Initial treatment with rectal diazepam was more common in those with seizure duration >30 min. Most children with PFS are treated with antiepileptic drugs early by the ambulance service. However, even timely treatment does not prevent status epilepticus in the majority of cases. These data highlight the need for effective early treatment of this common pediatric emergency.
Product Description
Supelco
Diazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Diazepam