Merck
  • Synthesis and spectroscopic properties of modular platinum(II) terpyridine phosphorescent pockets.
Synthesis and spectroscopic properties of modular platinum(II) terpyridine phosphorescent pockets.
Dalton transactions (Cambridge, England : 2003) (2009-06-11)
Wah-Leung Tong, Michael C W Chan, Nianyong Zhu, Sze-Kar Leung
ABSTRACT
A modular design approach has been utilized to develop molecular 'pockets' featuring three integrated components, namely a phosphorescent Pt(II)-pi(organic) moiety, a suitable receptor group, and a rigid, conjugated connecting backbone, and the cavities therein are examined by NMR spectroscopy and an X-ray crystal structure. Their photophysical properties have been investigated; unusually for Pt-terpyridine derivatives, the complexes are luminescent (near-red (3)MLCT emission) in solution at room temperature. It has been shown that their emissive characteristics are affected by the nature of the backbone component but not the receptor group.
Sigma-Aldrich
2,2′:6′,2′′-Terpyridine, 98%