Merck
NADH oxidase activity of indoleamine 2,3-dioxygenase.
The Journal of biological chemistry (2011-06-22)
Federico I Rosell, Hsin H Kuo, A Grant Mauk
ABSTRACT
The heme enzyme indoleamine 2,3-dioxygenase (IDO) was found to oxidize NADH under aerobic conditions in the absence of other enzymes or reactants. This reaction led to the formation of the dioxygen adduct of IDO and supported the oxidation of Trp to N-formylkynurenine. Formation of the dioxygen adduct and oxidation of Trp were accelerated by the addition of small amounts of hydrogen peroxide, and both processes were inhibited in the presence of either superoxide dismutase or catalase. Anaerobic reaction of IDO with NADH proceeded only in the presence of a mediator (e.g. methylene blue) and resulted in formation of the ferrous form of the enzyme. We propose that trace amounts of peroxide previously proposed to occur in NADH solutions as well as solid NADH activate IDO and lead to aerobic formation of superoxide and the reactive dioxygen adduct of the enzyme.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, ~98%, pkg of 100 mg
Pricing and availability is not currently available.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Pricing and availability is not currently available.