Merck
  • BEX2 suppresses mitochondrial activity and is required for dormant cancer stem cell maintenance in intrahepatic cholangiocarcinoma.
BEX2 suppresses mitochondrial activity and is required for dormant cancer stem cell maintenance in intrahepatic cholangiocarcinoma.
Scientific reports (2020-12-11)
Keiichi Tamai, Mao Nakamura-Shima, Rie Shibuya-Takahashi, Shin-Ichiro Kanno, Akira Yasui, Mai Mochizuki, Wataru Iwai, Yuta Wakui, Makoto Abue, Kuniharu Yamamoto, Koh Miura, Masamichi Mizuma, Michiaki Unno, Sadafumi Kawamura, Ikuro Sato, Jun Yasuda, Kazunori Yamaguchi, Kazuo Sugamura, Kennichi Satoh
ABSTRACT
Cancer stem cells (CSCs) define a subpopulation of cancer cells that are resistant to therapy. However, little is known of how CSC characteristics are regulated. We previously showed that dormant cancer stem cells are enriched with a CD274low fraction of cholangiocarcinoma cells. Here we found that BEX2 was highly expressed in CD274low cells, and that BEX2 knockdown decreased the tumorigenicity and G0 phase of cholangiocarcinoma cells. BEX2 was found to be expressed predominantly in G0 phase and starvation induced the USF2 transcriptional factor, which induced BEX2 transcription. Comprehensive screening of BEX2 binding proteins identified E3 ubiquitin ligase complex proteins, FEM1B and CUL2, and a mitochondrial protein TUFM, and further demonstrated that knockdown of BEX2 or TUFM increased mitochondria-related oxygen consumption and decreased tumorigenicity in cholangiocarcinoma cells. These results suggest that BEX2 is essential for maintaining dormant cancer stem cells through the suppression of mitochondrial activity in cholangiocarcinoma.
Sigma-Aldrich
MISSION® esiRNA, targeting human FEM1B
Sigma-Aldrich
MISSION® esiRNA, targeting human TUFM
Sigma-Aldrich
MISSION® esiRNA, targeting human IVD (1)
Sigma-Aldrich
MISSION® esiRNA, targeting human PECR