Merck
  • Wnt signaling mediates oncogenic synergy between Akt and Dlx5 in T-cell lymphomagenesis by enhancing cholesterol synthesis.
Wnt signaling mediates oncogenic synergy between Akt and Dlx5 in T-cell lymphomagenesis by enhancing cholesterol synthesis.
Scientific reports (2020-09-29)
Yinfei Tan, Eleonora Sementino, Zemin Liu, Kathy Q Cai, Joseph R Testa
ABSTRACT
The Dlx5 homeobox gene was first implicated as an oncogene in a T-ALL mouse model expressing myristoylated (Myr) Akt2. Furthermore, overexpression of Dlx5 was sufficient to drive T-ALL in mice by directly activating Akt and Notch signaling. These findings implied that Akt2 cooperates with Dlx5 in T-cell lymphomagenesis. To test this hypothesis, Lck-Dlx5;Lck-MyrAkt2 transgenic mice were generated. MyrAkt2 synergized with Dlx5 to greatly accelerate and enhance the dissemination of T-lymphomagenesis. RNA-seq analysis performed on lymphomas from Lck-Dlx5;Lck-MyrAkt mice revealed upregulation of genes involved in the Wnt and cholesterol biosynthesis pathways. Combined RNA-seq and ChIP-seq analysis of lymphomas from Lck-Dlx5;Lck-MyrAkt mice demonstrated that β-catenin directly regulates genes involved in sterol regulatory element binding transcription factor 2 (Srebf2)-cholesterol synthesis. These lymphoma cells had high Lef1 levels and were highly sensitive to β-catenin and Srebf2-cholesterol synthesis inhibitors. Similarly, human T-ALL cell lines with activated NOTCH and AKT and elevated LEF1 levels were sensitive to inhibition of β-catenin and cholesterol pathways. Furthermore, LEF1 expression positively correlated with expression of genes involved in the cholesterol synthesis pathway in primary human T-ALL specimens. Together, these data suggest that targeting β-catenin and/or cholesterol biosynthesis, together with AKT, could have therapeutic efficacy in a subset of T-ALL patients.
Sigma-Aldrich
OSC Inhibitor, Ro 48-8071, The OSC Inhibitor, Ro 48-8071, also referenced under CAS 189197-69-1, controls the biological activity of 2,3-oxidosqualene:lanosterol cyclase activity (OSC).
Sigma-Aldrich
β-Catenin/Tcf Inhibitor II, PKF118-310, The β-Catenin/Tcf Inhibitor II, PKF118-310 controls the biological activity of β-Catenin/Tcf. This small molecule/inhibitor is primarily used for Cancer applications.
Sigma-Aldrich
Simvastatin, Sodium Salt, InSolution, ≥95%