Merck
  • Occupational exposure to airborne formaldehyde in hospital: setting an automatic sampling system, comparing different monitoring methods and applying them to assess exposure.
Occupational exposure to airborne formaldehyde in hospital: setting an automatic sampling system, comparing different monitoring methods and applying them to assess exposure.
La Medicina del lavoro (2019-12-18)
Nicola Mucci, Stefano Dugheri, Venerando Rapisarda, Marcello Campagna, Giacomo Garzaro, Andrea Farioli, Giovanni Cappelli, Giulio Arcangeli
ABSTRACT
In recent years, under-vacuum sealing (UVS) and containers with formalin encapsulated in the lid have been proposed for the reduction of occupational exposure to airborne formaldehyde (FA) in healthcare environments. We are presenting a study focused on the assessment of FA in hospitals: an automatic sampling system was set, different sampling devices were compared, and the concentration of FA was assessed, following its use in different scenarios. Three different devices for sampling/measuring FA were compared. They are based on: 1. silica gel cartridges impregnated with 2,4-dinitrophenylhydrazine (2,4-DNPH); 2. SPME® fiber using O-(2,3,4,5,6-pentafluorobenzyl)hydroxylamine; 3. direct reading commercial instrumentation. Three typical scenarios using FA were investigated: operating theatres where small biopsies are soaked into closed-circuit system 4% FA containers, secretariat of pathology laboratories during the registration of biopsies and pathology laboratories during the filling procedure by UVS and the slicing of biopsies. The automatic sampling system allowed short-, long-, and in continuous-sampling time to measure airborne FA. Different sampling devices provided comparable results when tested to assess FA concentration ranging from 0.020-0.320 ppm in a test chamber, although  the devices based on 2,4-DNPH were  the best in terms of sensitivity and accuracy. The results of 246 samples showed that the FA concentration was less than 0.04 ppm in 91% of the measurements. The automatic methods efficiently allow sampling and measurement of FA in hospital settings. When using safe practices, the concentration of FA is well below occupational limit values.
Product Description
Supelco
Formaldehyde O-pentafluorophenylmethyl-oxime, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
25 mg
Fulfilment and delivery delayed
100 mg
Fulfilment and delivery delayed
500 mg
Fulfilment and delivery delayed
1 g
Fulfilment and delivery delayed
To order products, please contact your local dealer.
Supelco
Formaldehyde-2,4-DNPH, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
25 mg
Fulfilment and delivery delayed
100 mg
Fulfilment and delivery delayed
500 mg
Fulfilment and delivery delayed
1 g
Fulfilment and delivery delayed
To order products, please contact your local dealer.