Merck
  • Pro-inflammatory activation of microglia in the brain of patients with sepsis.
Pro-inflammatory activation of microglia in the brain of patients with sepsis.
Neuropathology and applied neurobiology (2018-05-29)
T Zrzavy, R Höftberger, T Berger, H Rauschka, O Butovsky, H Weiner, H Lassmann
ABSTRACT
Experimental data suggest that systemic immune activation may create a pro-inflammatory environment with microglia activation in the central nervous system in the absence of overt inflammation, which in turn may be deleterious in conditions of neurodegenerative disease. The extent to which this is relevant for the human brain is unknown. The central aim of this study is to provide an in-depth characterization of the microglia and macrophage response to systemic inflammation. We used recently described markers to characterize the origin and functional states of microglia/macrophages in white and grey matter in patients who died under septic conditions and compared it to those patients without systemic inflammation. We found pro-inflammatory microglia activation in septic patients in the white matter, with very little activation in the grey matter. Using a specific marker for resident microglia (TMEM119), we found that parenchyma microglia were activated and that there was additional recruitment of perivascular macrophages. Pro-inflammatory microglia activation occurred in the presence of homeostatic microglia cells. In contrast to inflammatory or ischaemic diseases of the brain, the anti-inflammatory microglia markers CD163 or CD206 were not expressed in acute sepsis. Furthermore, we found pronounced upregulation of inducible nitric oxide synthase not only in microglia, but also in astrocytes and endothelial cells. Our results demonstrate the pronounced effects of systemic inflammation on the human brain and have important implications for the selection of control populations for studies on microglia activation in human brain disease.
Sigma-Aldrich
Anti-TMEM119 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Ferritin, Human antibody produced in rabbit, whole antiserum, liquid