![Tiamulin for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/296/143/83d02d72-9bcd-4a27-8bc2-b52b73f64f5c/640/83d02d72-9bcd-4a27-8bc2-b52b73f64f5c.png)
Tiamulin for peak identification
![Tiamulin for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/296/143/83d02d72-9bcd-4a27-8bc2-b52b73f64f5c/640/83d02d72-9bcd-4a27-8bc2-b52b73f64f5c.png)
Synonym(s):
Tiamulin fumarate, Thiamutilin fumarate
Empirical Formula (Hill Notation):
C32H51NO8S
CAS Number:
Molecular Weight:
609.81
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing