Skip to Content
Merck
Perphenazine European Pharmacopoeia (EP) Reference Standard

Perphenazine

Perphenazine European Pharmacopoeia (EP) Reference Standard
Synonym(s):
4-[3-(2-Chloro-10H-phenothiazin-10-yl)propyl]-1-piperazineethanol, Perphenazine
Empirical Formula (Hill Notation):
C21H26ClN3OS
CAS Number:
Molecular Weight:
403.97

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade