
Peiminine

Synonym(s):
3β,20-Dihydroxy-5α-cevan-6-one, Fritillarine, Osnovanin, Raddeanine, Verticinone
Empirical Formula (Hill Notation):
C27H43NO3
CAS Number:
Molecular Weight:
429.64
Products
Formula Weight
Manufacturer
Product Line
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing