Skip to Content
Merck
Formaldehyde-2,4-DNPH certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Formaldehyde-2,4-DNPH

Formaldehyde-2,4-DNPH certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Formaldehyde-2,4-dinitrophenylhydrazone
Linear Formula:
H2C=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
210.15

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing