
Fluoxetine-D6 oxalate solution

Empirical Formula (Hill Notation):
C17D6H12F3NO · C2H2O4
Molecular Weight:
405.40
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing