
Famotidine impurity D

Synonym(s):
3-{{{2-[(Diaminomethylene)amino]thiazol-4-yl}methyl}sulphanyl}propanamide, 3-[[[2-[(Aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]propanamide
Empirical Formula (Hill Notation):
C8H13N5OS2
CAS Number:
Molecular Weight:
259.35
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing