
DMT-2′O-TBDMS-rG(ib) Phosphoramidite

Synonym(s):
N-(2-methyl-1-oxopropyl)-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-guanosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]
Empirical Formula (Hill Notation):
C50H68N7O9PSi
CAS Number:
Molecular Weight:
970.18
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-4 of 4