
Cyclohexyl methacrylate

Synonym(s):
Methacrylic acid cyclohexyl ester
Linear Formula:
CH2=C(CH3)COOC6H11
CAS Number:
Molecular Weight:
168.23
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Monomer Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing