
Cyclohexyl isocyanide

Synonym(s):
Isocyanocyclohexane, Cyclohexyl isocyanide
Linear Formula:
C6H11NC
CAS Number:
Molecular Weight:
109.17
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing