
Clobetasol propionate

Synonym(s):
Clobetasol propionate
Empirical Formula (Hill Notation):
C25H32ClFO5
CAS Number:
Molecular Weight:
466.97
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing