Skip to Content
Merck
Cimetidine Impurity I Pharmaceutical Secondary Standard; Certified Reference Material

Cimetidine Impurity I

Cimetidine Impurity I Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
5-Methyl-1H-imidazole-4-methanol, 4-(Hydroxymethyl)-5-methylimidazole
Empirical Formula (Hill Notation):
C5H8N2O
CAS Number:
Molecular Weight:
112.13

Products

Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1