
Chloroperoxidase from Caldariomyces fumago

Synonym(s):
Chloride Peroxidase, Chloride:hydrogen-peroxide oxidoreductase
CAS Number:
Products
Product Category
Brand
Biological Source
Color
Physical Form
Showing 1-3 of 3
Compare
Product Number
Description
Pricing