
Cefoxitin

Synonym(s):
(6R-cis)-3-[[(Aminocarbonyl)oxy]methyl]-7-methoxy-8-oxo-7-[(2-thienylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Empirical Formula (Hill Notation):
C16H17N3O7S2
CAS Number:
Molecular Weight:
427.45
Products
Brand
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2