![Benzyl mercaptan United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/237/249/80e252fa-7cb4-4c5d-babf-e018f2dc73cd/640/80e252fa-7cb4-4c5d-babf-e018f2dc73cd.png)
Benzyl mercaptan
![Benzyl mercaptan United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/237/249/80e252fa-7cb4-4c5d-babf-e018f2dc73cd/640/80e252fa-7cb4-4c5d-babf-e018f2dc73cd.png)
Synonym(s):
α-Toluenethiol, α-Tolyl mercaptan, Benzenemethanethiol
Linear Formula:
C6H5CH2SH
CAS Number:
Molecular Weight:
124.20
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing