
Benzatropine mesilate

Synonym(s):
Benztropine mesylate, Benzatropine mesylate, Benzotropine mesylate, Benztropine methanesulfonate
Empirical Formula (Hill Notation):
C21H25NO · CH3SO3H
CAS Number:
Molecular Weight:
403.53
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing