
ASTM Colour Reference Standard ASTM D 6045, D1500 (ASTM colour 5)

Synonym(s):
ASTM Colour Reference Standard
Products
Brand
Manufacturer
Agency
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing