
Anthralin

Synonym(s):
Dithranol, 1,8,9-Anthracenetriol, 1,8-Dihydroxy-9(10H)-anthracenone, 1,8-Dihydroxyanthrone, Anthralin
Empirical Formula (Hill Notation):
C14H10O3
CAS Number:
Molecular Weight:
226.23
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing