![Anhydrotetracycline hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/371/544/d0c1b194-324d-4c22-aebb-4a87aa7fa69a/640/d0c1b194-324d-4c22-aebb-4a87aa7fa69a.png)
Anhydrotetracycline hydrochloride
![Anhydrotetracycline hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/371/544/d0c1b194-324d-4c22-aebb-4a87aa7fa69a/640/d0c1b194-324d-4c22-aebb-4a87aa7fa69a.png)
Synonym(s):
(4S,4aS,12aS)-4-(Dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide monohydrochloride, Anhydrotetracycline hydrochloride
Empirical Formula (Hill Notation):
C22H22N2O7 · HCl
CAS Number:
Molecular Weight:
462.88
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing