
Actinomycin D, Ready Made Solution

Synonym(s):
Actinomycin D solution from Streptomyces sp., Actinomycin IV, Dactinomycin
Empirical Formula (Hill Notation):
C62H86N12O16
CAS Number:
Molecular Weight:
1255.42
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing