![Acetic acid – triethylamine solution 1:1 suitable for HPLC, 2M:2M aqueous solution, LiChropur™](/deepweb/assets/sigmaaldrich/product/structures/832/119/b228d74b-ad9f-45bb-9edd-c0e877526789/640/b228d74b-ad9f-45bb-9edd-c0e877526789.png)
Acetic acid – triethylamine solution 1:1
![Acetic acid – triethylamine solution 1:1 suitable for HPLC, 2M:2M aqueous solution, LiChropur™](/deepweb/assets/sigmaaldrich/product/structures/832/119/b228d74b-ad9f-45bb-9edd-c0e877526789/640/b228d74b-ad9f-45bb-9edd-c0e877526789.png)
Synonym(s):
Triethylamine : Acetic acid 1:1 solution, Triethylammonium acetate solution
Linear Formula:
CH3COOH · N(CH2CH3)3
CAS Number:
Molecular Weight:
161.24
Products
Product Category
Brand
Formula Weight
Physical Form
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing